RIE156326585 Rieke Metals, Inc.

4'-Bromo-2'-fluoro-3-phenylpropiophenone

  • MDL number MFCD03842945
  • Cas Number : 898764-54-0
  • Molecular Weight : 307.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromo-2-fluorophenyl)-3-phenyl-1-propanone
Inchi Code 1S/C15H12BrFO/c16-12-7-8-13(14(17)10-12)15(18)9-6-11-4-2-1-3-5-11/h1-5,7-8,10H,6,9H2
Inchi Key WZFGKUHXSCEYTQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0100-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-49
Availability details for
Quantity
  • Availability
  • Add to Cart