RIE156326588 Rieke Metals, Inc.

4'-chloro-2'-fluoro-3-phenylpropiophenone

  • MDL number MFCD03842948
  • Cas Number : 674767-23-8
  • Molecular Weight : 262.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0100-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-52
Availability details for
Quantity
  • Availability
  • Add to Cart