RIE156326593 Rieke Metals, Inc.

3',5'-dichloro-3-phenylpropiophenone

  • MDL number MFCD03842953
  • Cas Number : 898788-84-6
  • Molecular Weight : 279.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0100-57

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-57
Availability details for
Quantity
  • Availability
  • Add to Cart