RIE156326594 Rieke Metals, Inc.

2',4'-difluoro-3-phenylpropiophenone

  • MDL number MFCD03842954
  • Cas Number : 898788-87-9
  • Molecular Weight : 246.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,4-difluorophenyl)-3-phenyl-1-propanone
Inchi Code 1S/C15H12F2O/c16-12-7-8-13(14(17)10-12)15(18)9-6-11-4-2-1-3-5-11/h1-5,7-8,10H,6,9H2
Inchi Key YDXVCFGAFOMMFK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0100-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-58
Availability details for
Quantity
  • Availability
  • Add to Cart