RIE156326595 Rieke Metals, Inc.

3',4'-difluoro-3-phenylpropiophenone

  • MDL number MFCD03842955
  • Cas Number : 898788-90-4
  • Molecular Weight : 246.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0100-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0100-59
Availability details for
Quantity
  • Availability
  • Add to Cart