RIE156326600 Rieke Metals, Inc.

cyclobutyl 2-phenethyl ketone

  • MDL number MFCD03842959
  • Cas Number : 874009-12-8
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclobutyl-3-phenyl-1-propanone
Inchi Code 1S/C13H16O/c14-13(12-7-4-8-12)10-9-11-5-2-1-3-6-11/h1-3,5-6,12H,4,7-10H2
Inchi Key RNUNGNSPAYQLSI-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0102-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0102-00
Availability details for
Quantity
  • Availability
  • Add to Cart