RIE156326604 Rieke Metals, Inc.

4'-methyl-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842964
  • Cas Number : 898789-10-1
  • Molecular Weight : 238.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0200-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-04
Availability details for
Quantity
  • Availability
  • Add to Cart