RIE156326606 Rieke Metals, Inc.

3'-methoxy-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842966
  • Cas Number : 898789-16-7
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0200-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-06
Availability details for
Quantity
  • Availability
  • Add to Cart