RIE156326610 Rieke Metals, Inc.

4'-cyano-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842970
  • Cas Number : 898789-26-9
  • Molecular Weight : 249.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0200-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-10
Availability details for
Quantity
  • Availability
  • Add to Cart