RIE156326612 Rieke Metals, Inc.

3'-carboethoxy-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842972
  • Cas Number : 898789-30-5
  • Molecular Weight : 296.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-[3-(2-methylphenyl)propanoyl]benzoate
Inchi Code 1S/C19H20O3/c1-3-22-19(21)17-10-6-9-16(13-17)18(20)12-11-15-8-5-4-7-14(15)2/h4-10,13H,3,11-12H2,1-2H3
Inchi Key AXGYEASTDYZSMV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0200-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-12
Availability details for
Quantity
  • Availability
  • Add to Cart