RIE156326618 Rieke Metals, Inc.

3'-chloro-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842978
  • Cas Number : 898789-42-9
  • Molecular Weight : 258.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-chlorophenyl)-3-(2-methylphenyl)-1-propanone
Inchi Code 1S/C16H15ClO/c1-12-5-2-3-6-13(12)9-10-16(18)14-7-4-8-15(17)11-14/h2-8,11H,9-10H2,1H3
Inchi Key OOIMKZCIDSQENR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0200-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-20
Availability details for
Quantity
  • Availability
  • Add to Cart