RIE156326625 Rieke Metals, Inc.

2',6'-dimethyl-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842985
  • Cas Number : 898789-56-5
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2,6-dimethylphenyl)-3-(2-methylphenyl)-1-propanone
Inchi Code 1S/C18H20O/c1-13-7-4-5-10-16(13)11-12-17(19)18-14(2)8-6-9-15(18)3/h4-10H,11-12H2,1-3H3
Inchi Key MYVIOXDAYLMPFQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0200-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-29
Availability details for
Quantity
  • Availability
  • Add to Cart