RIE156326627 Rieke Metals, Inc.

3',5'-dimethyl-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842987
  • Cas Number : 898789-60-1
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,5-dimethylphenyl)-3-(2-methylphenyl)-1-propanone
Inchi Code 1S/C18H20O/c1-13-10-14(2)12-17(11-13)18(19)9-8-16-7-5-4-6-15(16)3/h4-7,10-12H,8-9H2,1-3H3
Inchi Key YIHJRQPSNGVJIU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0200-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-31
Availability details for
Quantity
  • Availability
  • Add to Cart