RIE156326630 Rieke Metals, Inc.

3'-chloro-4'-fluoro-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842990
  • Cas Number : 898789-66-7
  • Molecular Weight : 276.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-chloro-4-fluorophenyl)-3-(2-methylphenyl)-1-propanone
Inchi Code 1S/C16H14ClFO/c1-11-4-2-3-5-12(11)7-9-16(19)13-6-8-15(18)14(17)10-13/h2-6,8,10H,7,9H2,1H3
Inchi Key BVBYNTLPTXOINS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0200-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-34
Availability details for
Quantity
  • Availability
  • Add to Cart