RIE156326632 Rieke Metals, Inc.

2'-fluoro-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842992
  • Cas Number : 898789-70-3
  • Molecular Weight : 242.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0200-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-37
Availability details for
Quantity
  • Availability
  • Add to Cart