RIE156326633 Rieke Metals, Inc.

3-(2-methylphenyl)-2'-trifluoromethylpropiophenone

  • MDL number MFCD03842993
  • Cas Number : 898789-73-6
  • Molecular Weight : 292.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0200-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-43
Availability details for
Quantity
  • Availability
  • Add to Cart