RIE156326635 Rieke Metals, Inc.

3-(2-methylphenyl)-4'-trifluoromethylpropiophenone

  • MDL number MFCD03842995
  • Cas Number : 898789-79-2
  • Molecular Weight : 292.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0200-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-45
Availability details for
Quantity
  • Availability
  • Add to Cart