RIE156326636 Rieke Metals, Inc.

4'-bromo-2'-fluoro-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842996
  • Cas Number : 898789-82-7
  • Molecular Weight : 321.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0200-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-49
Availability details for
Quantity
  • Availability
  • Add to Cart