RIE156326638 Rieke Metals, Inc.

3'-chloro-5'-fluoro-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842998
  • Cas Number : 898789-88-3
  • Molecular Weight : 276.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0200-51

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-51
Availability details for
Quantity
  • Availability
  • Add to Cart