RIE156326639 Rieke Metals, Inc.

4'-chloro-2'-fluoro-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03842999
  • Cas Number : 898789-91-8
  • Molecular Weight : 276.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-chloro-2-fluorophenyl)-3-(2-methylphenyl)-1-propanone
Inchi Code 1S/C16H14ClFO/c1-11-4-2-3-5-12(11)6-9-16(19)14-8-7-13(17)10-15(14)18/h2-5,7-8,10H,6,9H2,1H3
Inchi Key LLPWLLUIMACHQD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0200-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-52
Availability details for
Quantity
  • Availability
  • Add to Cart