RIE156326641 Rieke Metals, Inc.

2',4'-dichloro-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03843001
  • Cas Number : 898789-96-3
  • Molecular Weight : 293.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0200-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-54
Availability details for
Quantity
  • Availability
  • Add to Cart