RIE156326646 Rieke Metals, Inc.

3',4'-difluoro-3-(2-methylphenyl)propiophenone

  • MDL number MFCD03843006
  • Cas Number : 898790-11-9
  • Molecular Weight : 260.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,4-difluorophenyl)-3-(2-methylphenyl)-1-propanone
Inchi Code 1S/C16H14F2O/c1-11-4-2-3-5-12(11)7-9-16(19)13-6-8-14(17)15(18)10-13/h2-6,8,10H,7,9H2,1H3
Inchi Key PMHYNFUFHNOMLH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0200-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-59
Availability details for
Quantity
  • Availability
  • Add to Cart