RIE156326648 Rieke Metals, Inc.

3-(2-methylphenyl)-3',4',5'-trifluoropropiophenone

  • MDL number MFCD03843008
  • Cas Number : 898790-17-5
  • Molecular Weight : 278.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-methylphenyl)-1-(3,4,5-trifluorophenyl)-1-propanone
Inchi Code 1S/C16H13F3O/c1-10-4-2-3-5-11(10)6-7-15(20)12-8-13(17)16(19)14(18)9-12/h2-5,8-9H,6-7H2,1H3
Inchi Key KQRNMRKURLYFAL-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0200-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0200-61
Availability details for
Quantity
  • Availability
  • Add to Cart