RIE156326650 Rieke Metals, Inc.

cyclopropyl 2-(2-methylphenyl)ethyl ketone

  • MDL number MFCD03843009
  • Cas Number : 898790-23-3
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0201-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0201-00
Availability details for
Quantity
  • Availability
  • Add to Cart