RIE156326652 Rieke Metals, Inc.

cyclopentyl 2-(2-methylphenyl)ethyl ketone

  • MDL number MFCD03843011
  • Cas Number : 898790-27-7
  • Molecular Weight : 216.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0203-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0203-00
Availability details for
Quantity
  • Availability
  • Add to Cart