RIE156326653 Rieke Metals, Inc.

cyclohexyl 2-(2-methylphenyl)ethyl ketone

  • MDL number MFCD03843012
  • Cas Number : 898790-29-9
  • Molecular Weight : 230.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclohexyl-3-(2-methylphenyl)-1-propanone
Inchi Code 1S/C16H22O/c1-13-7-5-6-8-14(13)11-12-16(17)15-9-3-2-4-10-15/h5-8,15H,2-4,9-12H2,1H3
Inchi Key QTQMWLBLIXRJSW-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0204-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0204-00
Availability details for
Quantity
  • Availability
  • Add to Cart