RIE156326654 Rieke Metals, Inc.

3-(3-methylphenyl)propiophenone

  • MDL number MFCD03843013
  • Cas Number : 95465-70-6
  • Molecular Weight : 224.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0300-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-01
Availability details for
Quantity
  • Availability
  • Add to Cart