RIE156326659 Rieke Metals, Inc.

3'-methoxy-3-(3-methylphenyl)propiophenone

  • MDL number MFCD03843018
  • Cas Number : 898790-39-1
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0300-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-06
Availability details for
Quantity
  • Availability
  • Add to Cart