RIE156326667 Rieke Metals, Inc.

3-(3-methylphenyl)-2'-thiomethylpropiophenone

  • MDL number MFCD07699573
  • Cas Number : 898790-55-1
  • Molecular Weight : 270.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3-methylphenyl)-1-[2-(methylsulfanyl)phenyl]-1-propanone
Inchi Code 1S/C17H18OS/c1-13-6-5-7-14(12-13)10-11-16(18)15-8-3-4-9-17(15)19-2/h3-9,12H,10-11H2,1-2H3
Inchi Key UWQCFPQILHBLEY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0300-14

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-14
Availability details for
Quantity
  • Availability
  • Add to Cart