RIE156326668 Rieke Metals, Inc.

3-(3-methylphenyl)-4'-thiomethylpropiophenone

  • MDL number MFCD07699574
  • Cas Number : 898790-57-3
  • Molecular Weight : 270.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0300-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-15
Availability details for
Quantity
  • Availability
  • Add to Cart