RIE156326669 Rieke Metals, Inc.

3'-bromo-3-(3-methylphenyl)propiophenone

  • MDL number MFCD07699575
  • Cas Number : 898790-59-5
  • Molecular Weight : 303.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0300-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-18
Availability details for
Quantity
  • Availability
  • Add to Cart