RIE156326671 Rieke Metals, Inc.

3'-chloro-3-(3-methylphenyl)propiophenone

  • MDL number MFCD07699577
  • Cas Number : 898790-63-1
  • Molecular Weight : 258.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-chlorophenyl)-3-(3-methylphenyl)-1-propanone
Inchi Code 1S/C16H15ClO/c1-12-4-2-5-13(10-12)8-9-16(18)14-6-3-7-15(17)11-14/h2-7,10-11H,8-9H2,1H3
Inchi Key ZZEPFYOPBKHWMX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0300-20

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-20
Availability details for
Quantity
  • Availability
  • Add to Cart