RIE156326673 Rieke Metals, Inc.

3'-fluoro-3-(3-methylphenyl)propiophenone

  • MDL number MFCD07699579
  • Cas Number : 898790-67-5
  • Molecular Weight : 242.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0300-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-22
Availability details for
Quantity
  • Availability
  • Add to Cart