RIE156326675 Rieke Metals, Inc.

2',3'-dimethyl-3-(3-methylphenyl)propiophenone

  • MDL number MFCD07699581
  • Cas Number : 898790-69-7
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0300-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-26
Availability details for
Quantity
  • Availability
  • Add to Cart