RIE156326681 Rieke Metals, Inc.

4'-bromo-3'-fluoro-3-(3-methylphenyl)propiophenone

  • MDL number MFCD07699587
  • Cas Number : 898790-87-9
  • Molecular Weight : 321.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0300-32

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-32
Availability details for
Quantity
  • Availability
  • Add to Cart