RIE156326687 Rieke Metals, Inc.

3-(3-methylphenyl)-3'-trifluoromethylpropiophenone

  • MDL number MFCD07699593
  • Cas Number : 898791-05-4
  • Molecular Weight : 292.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0300-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-44
Availability details for
Quantity
  • Availability
  • Add to Cart