RIE156326690 Rieke Metals, Inc.

2'-chloro-4'-fluoro-3-(3-methylphenyl)propiophenone

  • MDL number MFCD07699596
  • Cas Number : 898791-14-5
  • Molecular Weight : 276.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0300-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-50
Availability details for
Quantity
  • Availability
  • Add to Cart