RIE156326693 Rieke Metals, Inc.

2',3'-dichloro-3-(3-methylphenyl)propiophenone

  • MDL number MFCD07699599
  • Cas Number : 898791-23-6
  • Molecular Weight : 293.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0300-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-53
Availability details for
Quantity
  • Availability
  • Add to Cart