RIE156326698 Rieke Metals, Inc.

2',4'-difluoro-3-(3-methylphenyl)propiophenone

  • MDL number MFCD07699604
  • Cas Number : 898768-16-6
  • Molecular Weight : 260.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0300-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0300-58
Availability details for
Quantity
  • Availability
  • Add to Cart