RIE156326703 Rieke Metals, Inc.

cyclopropyl 2-(3-methylphenyl)ethyl ketone

  • MDL number MFCD03843025
  • Cas Number : 898768-30-4
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0301-00

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0301-00
Availability details for
Quantity
  • Availability
  • Add to Cart