RIE156326708 Rieke Metals, Inc.

3'-methyl-3-(4-methylphenyl)propiophenone

  • MDL number MFCD03843031
  • Cas Number : 898768-45-1
  • Molecular Weight : 238.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0400-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-03
Availability details for
Quantity
  • Availability
  • Add to Cart