RIE156326709 Rieke Metals, Inc.

4'-methyl-3-(4-methylphenyl)propiophenone

  • MDL number MFCD03843032
  • Cas Number : 20615-46-7
  • Molecular Weight : 238.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0400-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-04
Availability details for
Quantity
  • Availability
  • Add to Cart