RIE156326710 Rieke Metals, Inc.

2'-methoxy-3-(4-methylphenyl)propiophenone

  • MDL number MFCD03843033
  • Cas Number : 898768-48-4
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0400-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-05
Availability details for
Quantity
  • Availability
  • Add to Cart