RIE156326712 Rieke Metals, Inc.

4'-Methoxy-3-(4-methylphenyl)propiophenone

  • MDL number MFCD03843035
  • Cas Number : 106511-65-3
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0400-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-07
Availability details for
Quantity
  • Availability
  • Add to Cart