RIE156326714 Rieke Metals, Inc.

3'-cyano-3-(4-methylphenyl)propiophenone

  • MDL number MFCD03843037
  • Cas Number : 898768-55-3
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[3-(4-methylphenyl)propanoyl]benzonitrile
Inchi Code 1S/C17H15NO/c1-13-5-7-14(8-6-13)9-10-17(19)16-4-2-3-15(11-16)12-18/h2-8,11H,9-10H2,1H3
Inchi Key WQXDIRFAWSJZST-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0400-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-09
Availability details for
Quantity
  • Availability
  • Add to Cart