RIE156326715 Rieke Metals, Inc.

4'-cyano-3-(4-methylphenyl)propiophenone

  • MDL number MFCD03843038
  • Cas Number : 898768-57-5
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[3-(4-methylphenyl)propanoyl]benzonitrile
Inchi Code 1S/C17H15NO/c1-13-2-4-14(5-3-13)8-11-17(19)16-9-6-15(12-18)7-10-16/h2-7,9-10H,8,11H2,1H3
Inchi Key YLJADAJAHOTNPH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0400-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-10
Availability details for
Quantity
  • Availability
  • Add to Cart