RIE156326716 Rieke Metals, Inc.

2'-carboethoxy-3-(4-methylphenyl)propiophenone

  • MDL number MFCD03843039
  • Cas Number : 898768-59-7
  • Molecular Weight : 296.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0400-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-11
Availability details for
Quantity
  • Availability
  • Add to Cart