RIE156326717 Rieke Metals, Inc.

3'-carboethoxy-3-(4-methylphenyl)propiophenone

  • MDL number MFCD07699608
  • Cas Number : 898768-61-1
  • Molecular Weight : 296.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-[3-(4-methylphenyl)propanoyl]benzoate
Inchi Code 1S/C19H20O3/c1-3-22-19(21)17-6-4-5-16(13-17)18(20)12-11-15-9-7-14(2)8-10-15/h4-10,13H,3,11-12H2,1-2H3
Inchi Key QYYJGGNRYLUROP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0400-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-12
Availability details for
Quantity
  • Availability
  • Add to Cart