RIE156326721 Rieke Metals, Inc.

3'-bromo-3-(4-methylphenyl)propiophenone

  • MDL number MFCD07699612
  • Cas Number : 898768-69-9
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-bromophenyl)-3-(4-methylphenyl)-1-propanone
Inchi Code 1S/C16H15BrO/c1-12-5-7-13(8-6-12)9-10-16(18)14-3-2-4-15(17)11-14/h2-8,11H,9-10H2,1H3
Inchi Key IULVLGKQGGBUOS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6127-0400-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-18
Availability details for
Quantity
  • Availability
  • Add to Cart