RIE156326732 Rieke Metals, Inc.

3',5'-dimethyl-3-(4-methylphenyl)propiophenone

  • MDL number MFCD07699623
  • Cas Number : 898768-87-1
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6127-0400-31

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6127-0400-31
Availability details for
Quantity
  • Availability
  • Add to Cart